Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166457.1 | NP_001159929.1 | 6 - 7 | 588 | 103 |
NM_001166459.1 | NP_001159931.1 | 6 - 7 | 588 | 103 | |
NM_145956.4 | NP_666068.1 | 6 - 7 | 588 | 103 | |
GenBank mRNA | AK011876.1 | - | 6 - 7 | 505 | 103 |
AK028483.1 | - | 4 - 5 | 2070 | 103 | |
AK030736.1 | - | 6 - 7 | 518 | 103 | |
AK038919.1 | - | 6 - 7 | 506 | 103 | |
AK149844.1 | - | 6 - 7 | 520 | 103 | |
AK164578.1 | - | 2 - 3 | 1671 | 103 | |
BC021313.1 | - | 6 - 7 | 535 | 103 | |
BC048179.1 | - | 6 - 7 | 502 | 103 | |
S68022.1 | - | 6 - 7 | 530 | 103 |