Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001080927.2 | NP_001074396.1 | 7 - 8 | 964 | 77 |
NM_001080928.1 | NP_001074397.1 | 7 - 8 | 850 | 77 | |
NM_001277116.1 | NP_001264045.1 | 9 - 10 | 1046 | 77 | |
NM_009035.5 | NP_033061.3 | 7 - 8 | 866 | 77 | |
GenBank mRNA | AK028363.1 | - | 9 - 10 | 1024 | 77 |
AK080359.1 | - | 7 - 8 | 759 | 77 | |
AK090006.1 | - | 7 - 8 | 751 | 77 | |
AK132145.1 | - | 7 - 8 | 805 | 77 | |
AK139087.1 | - | 7 - 8 | 751 | 77 | |
AK145187.1 | - | 7 - 8 | 758 | 77 | |
AK152746.1 | - | 7 - 8 | 866 | 77 | |
AK153163.1 | - | 8 - 9 | 754 | 77 | |
BC035299.1 | - | 5 - 6 | 652 | 77 | |
BC051387.1 | - | 7 - 8 | 795 | 77 | |
S63463.1 | - | 7 - 8 | 867 | 77 | |
X17459.1 | - | 7 - 8 | 862 | 77 |