Assay Design



Gene Transcripts

Gene Symbol
Sap18
Entrez Gene ID
20220
Gene Name
Sin3-associated polypeptide 18
Gene Aliases
C530046K05Rik, D11Ertd539e, EMegR4, Sinbp1
Chromosome Location
Chr.14: 57798189 - 57804980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009119.3 NP_033145.2 4 - 4 3314 103
GenBank mRNA BM899282.1 - 1 - 1 103
Z97062.1 - 4 - 4 3249 103

View Full Product Details