Assay Design



Gene Transcripts

Gene Symbol
Mcm3
Entrez Gene ID
17215
Gene Name
minichromosome maintenance deficient 3 (S. cerevisiae)
Gene Aliases
AL033361, C80350, Mcmd, P1, p1.m
Chromosome Location
Chr.1: 20803014 - 20820213 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008563.2 NP_032589.1 16 - 17 2240 60
GenBank mRNA AK088142.1 - 15 - 16 2746 60
AK133850.1 - 16 - 17 2305 60
AK145563.1 - 16 - 17 2339 60
AK146497.1 - 16 - 17 2253 60
AK147065.1 - 16 - 17 2255 60
AK148006.1 - 16 - 17 2285 60
BC020400.1 - 16 - 17 2255 60
BC031700.1 - 16 - 17 2240 60
X62154.1 - 15 - 16 2184 60

View Full Product Details