Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301737.1 | NP_001288666.1 | 19 - 20 | 2197 | 65 |
NM_010065.3 | NP_034195.2 | 18 - 19 | 2185 | 65 | |
NR_125959.1 | 19 - 20 | 2197 | 65 | ||
GenBank mRNA | AK144142.1 | - | 18 - 19 | 2136 | 65 |
AK220483.1 | - | 19 - 20 | 2139 | 65 | |
BC034679.1 | - | 19 - 20 | 2151 | 65 | |
BM950315.1 | - | 5 - 6 | 590 | 65 | |
CO425806.1 | - | 5 - 6 | 516 | 65 | |
L29457.1 | - | 3 - 4 | 223 | 65 | |
L31395.1 | - | 19 - 20 | 2094 | 65 | |
L31396.1 | - | 14 - 15 | 1366 | 65 | |
L31397.1 | - | 18 - 19 | 2138 | 65 |