Assay Design



Gene Transcripts

Gene Symbol
Asns
Entrez Gene ID
27053
Gene Name
asparagine synthetase
Gene Aliases
-
Chromosome Location
Chr.6: 7675165 - 7693182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012055.3 NP_036185.1 6 - 7 1005 68
GenBank mRNA AK076207.1 - 6 - 7 1023 68
AK160459.1 - 6 - 7 1020 68
AK167524.1 - 6 - 7 985 68
AK167933.1 - 6 - 7 1004 68
BC005552.1 - 6 - 7 1003 68
BC050123.1 - - 3333 68
U38940.1 - 6 - 7 1006 68

View Full Product Details