Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285870.1 | NP_001272799.1 | 10 - 11 | 2175 | 101 |
NM_001285871.1 | NP_001272800.1 | 10 - 11 | 2360 | 101 | |
NM_001285873.1 | NP_001272802.1 | 10 - 11 | 2175 | 101 | |
NM_018880.3 | NP_061368.1 | 10 - 11 | 2149 | 101 | |
GenBank mRNA | AB030912.1 | - | 10 - 11 | 2176 | 101 |
AF220019.1 | - | 10 - 11 | 2068 | 101 | |
AK019165.1 | - | 10 - 11 | 2110 | 101 | |
AK144674.1 | - | 10 - 11 | 2078 | 101 | |
AK170029.1 | - | 10 - 11 | 2113 | 101 | |
AK182206.1 | - | 1 - 2 | 88 | 101 | |
BC003347.1 | - | 10 - 11 | 2189 | 101 | |
BC034263.1 | - | 10 - 11 | 2114 | 101 |