Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020601.2 | NP_065626.1 | 3 - 4 | 249 | 73 |
GenBank mRNA | AK030547.1 | - | 3 - 4 | 464 | 73 |
AK036996.1 | - | 4 - 5 | 328 | 73 | |
AK038674.1 | - | 3 - 4 | 250 | 73 | |
BC043105.1 | - | 3 - 4 | 372 | 73 |