Assay Design



Gene Transcripts

Gene Symbol
Tulp1
Entrez Gene ID
22157
Gene Name
tubby like protein 1
Gene Aliases
Tulp1l
Chromosome Location
Chr.17: 28351515 - 28365211 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_021478.1 NP_067453.1 3 - 4 214 53
GenBank mRNA AF085681.1 - 3 - 4 262 53
AF105711.1 - 3 - 4 262 53
BC014820.1 - 3 - 4 219 53
BC029222.1 - 3 - 4 253 53

View Full Product Details