Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001162858.1 | NP_001156330.1 | 4 - 5 | 892 | 67 |
NM_021717.2 | NP_068363.2 | 4 - 5 | 892 | 67 | |
GenBank mRNA | AJ278170.1 | - | 4 - 5 | 915 | 67 |
AK134689.1 | - | 4 - 5 | 689 | 67 | |
AK156526.1 | - | 4 - 5 | 765 | 67 | |
BC094230.1 | - | 3 - 4 | 2015 | 67 | |
BC115523.1 | - | 4 - 5 | 642 | 67 |