Assay Design



Gene Transcripts

Gene Symbol
Coq9
Entrez Gene ID
67914
Gene Name
coenzyme Q9 homolog (yeast)
Gene Aliases
2310005O14Rik, C78387
Chromosome Location
Chr.8: 94838417 - 94854895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026452.2 NP_080728.1 2 - 3 232 93
GenBank mRNA AK009179.1 - 2 - 3 245 93
AK154343.1 - 2 - 3 257 93
AK158979.1 - 2 - 3 251 93
AK200178.1 - 1 - 2 122 93
BC036386.1 - 2 - 3 232 93

View Full Product Details