Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028339.1 | NP_082615.1 | 4 - 5 | 456 | 145 |
GenBank mRNA | AK013150.1 | - | 4 - 5 | 456 | 145 |
AK030429.1 | - | 4 - 5 | 474 | 145 | |
AK050567.1 | - | 4 - 5 | 455 | 145 | |
AK137059.1 | - | 4 - 5 | 453 | 145 | |
AK150511.1 | - | 4 - 5 | 463 | 145 | |
AK153027.1 | - | 4 - 5 | 459 | 145 | |
AK170468.1 | - | 4 - 5 | 474 | 145 | |
BC017603.1 | - | 4 - 5 | 456 | 145 | |
BC021533.1 | - | 4 - 5 | 477 | 145 |