Assay Design



Gene Transcripts

Gene Symbol
C8b
Entrez Gene ID
110382
Gene Name
complement component 8, beta polypeptide
Gene Aliases
4930439B20Rik, AI595927
Chromosome Location
Chr.4: 104764576 - 104804549 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133882.2 NP_598643.1 1 - 2 172 77
GenBank mRNA AB077304.1 - 1 - 2 94 77
AB077305.1 - 1 - 2 94 77
AB077306.1 - 1 - 2 94 77
AK050313.1 - 1 - 2 173 77
BC096382.1 - 1 - 2 146 77

View Full Product Details