Assay Design



Gene Transcripts

Gene Symbol
Kdm4a
Entrez Gene ID
230674
Gene Name
lysine (K)-specific demethylase 4A
Gene Aliases
D4Ertd222e, JHDM3A, Jmjd2, Jmjd2a, mKIAA0677
Chromosome Location
Chr.4: 118136957 - 118180043 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161823.1 NP_001155295.1 13 - 14 2236 71
NM_172382.2 NP_759014.2 13 - 14 2236 71
GenBank mRNA AK054095.1 - 13 - 14 2200 71
AK129187.1 - 13 - 14 2091 71
AK136085.1 - 4 - 5 893 71
AK145066.1 - 13 - 14 2222 71
AK145947.1 - 13 - 14 2158 71
BC028866.1 - 13 - 14 2139 71

View Full Product Details