Assay Design



Gene Transcripts

Gene Symbol
Mamdc2
Entrez Gene ID
71738
Gene Name
MAM domain containing 2
Gene Aliases
1200015L10Rik, AI591564, mamcan
Chromosome Location
Chr.19: 23302609 - 23448500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174857.3 NP_777282.1 5 - 6 911 68
GenBank mRNA AK035566.1 - 5 - 6 912 68
AK154219.1 - 5 - 6 988 68
BC042773.1 - 5 - 6 940 68

View Full Product Details