Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181397.2 | NP_852062.1 | 7 - 8 | 1436 | 72 |
GenBank mRNA | AK082517.1 | - | - | 1049 | 72 |
AK087803.1 | - | - | 1210 | 72 | |
AK088466.1 | - | 5 - 6 | 973 | 72 | |
AK152130.1 | - | 7 - 8 | 1358 | 72 | |
AK153288.1 | - | 7 - 8 | 1358 | 72 | |
AK155870.1 | - | 7 - 8 | 1400 | 72 | |
AK172884.1 | - | 7 - 8 | 1364 | 72 | |
BC043324.1 | - | 3 - 4 | 493 | 72 | |
BC052074.1 | - | 7 - 8 | 1436 | 72 |