Assay Design



Gene Transcripts

Gene Symbol
Rftn1
Entrez Gene ID
76438
Gene Name
raftlin lipid raft linker 1
Gene Aliases
2310015N21Rik, AI853526
Chromosome Location
Chr.17: 49993258 - 50190497 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181397.2 NP_852062.1 9 - 10 1630 78
GenBank mRNA AK082517.1 - - 1243 78
AK087803.1 - - 1404 78
AK088466.1 - 7 - 8 1167 78
AK152130.1 - 9 - 10 1552 78
AK153288.1 - 9 - 10 1552 78
AK155870.1 - 9 - 10 1594 78
AK172884.1 - 9 - 10 1558 78
BC043324.1 - 5 - 6 687 78
BC052074.1 - 9 - 10 1630 78

View Full Product Details