Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182959.3 | NP_892004.1 | 8 - 9 | 1444 | 80 |
GenBank mRNA | AF510321.1 | - | 8 - 9 | 1115 | 80 |
AK082743.1 | - | 8 - 9 | 1192 | 80 | |
AK149687.1 | - | 6 - 7 | 847 | 80 | |
BC042593.1 | - | 8 - 9 | 1459 | 80 |