Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_182959.3 | NP_892004.1 | 9 - 10 | 1575 | 70 |
GenBank mRNA | AF510321.1 | - | 9 - 10 | 1246 | 70 |
AK082743.1 | - | 9 - 10 | 1323 | 70 | |
AK149687.1 | - | 7 - 8 | 978 | 70 | |
BC042593.1 | - | 9 - 10 | 1590 | 70 |