Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_183019.2 | NP_898840.2 | 6 - 7 | 1182 | 60 |
GenBank mRNA | AK034013.1 | - | 6 - 7 | 1146 | 60 |
AK122448.2 | - | 2 - 3 | 237 | 60 | |
AK134629.1 | - | - | 1145 | 60 | |
AK138598.1 | - | 6 - 7 | 1142 | 60 | |
AK163424.1 | - | 6 - 7 | 1164 | 60 | |
AK169957.1 | - | 6 - 7 | 1202 | 60 | |
BC055804.1 | - | 6 - 7 | 1143 | 60 | |
BC094608.1 | - | - | 4702 | 60 | |
BC139371.1 | - | 6 - 7 | 1165 | 60 | |
BC139397.1 | - | 6 - 7 | 1165 | 60 |