Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001110228.1 | NP_001103698.1 | 14 - 14 | 7494 | 127 |
NM_001110229.1 | NP_001103699.1 | 15 - 15 | 7593 | 127 | |
NM_001110230.1 | NP_001103700.1 | 15 - 15 | 7581 | 127 | |
NM_001110231.1 | NP_001103701.1 | 16 - 16 | 8940 | 127 | |
NM_001110232.1 | NP_001103702.1 | 16 - 16 | 7567 | 127 | |
NM_001160292.1 | NP_001153764.1 | 14 - 14 | 7437 | 127 | |
NM_001160293.1 | NP_001153765.1 | 14 - 14 | 8598 | 127 | |
NM_010160.2 | NP_034290.2 | 12 - 12 | 8716 | 127 | |
GenBank mRNA | AK045654.1 | - | 1 - 1 | 1633 | 127 |
BC033049.1 | - | 1 - 1 | 730 | 127 |