Assay Design
Gene Transcripts
- Gene Symbol
- Mt2
- Entrez Gene ID
-
17750
- Gene Name
- metallothionein 2
- Gene Aliases
-
AA409533, MT-II, Mt-2
- Chromosome Location
-
Chr.8: 94172618 - 94173567 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_008630.2 |
NP_032656.1 |
3 - 3 |
495 |
111 |
GenBank mRNA |
AK002567.1 |
- |
3 - 3 |
330 |
111 |
View Full Product Details