Assay Design



Gene Transcripts

Gene Symbol
Emilin3
Entrez Gene ID
280635
Gene Name
elastin microfibril interfacer 3
Gene Aliases
1110013O17Rik, EMILIN-3, EMILIN-5, EMILIN-T, Emilin5
Chromosome Location
Chr.2: 160906438 - 160912339 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_182840.2 NP_878260.1 3 - 4 684 80
GenBank mRNA BC054738.1 - 3 - 4 673 80
BC057619.1 - 3 - 4 673 80

View Full Product Details