Assay Design



Gene Transcripts

Gene Symbol
Accs
Entrez Gene ID
329470
Gene Name
1-aminocyclopropane-1-carboxylate synthase (non-functional)
Gene Aliases
2610203E10Rik, Phacs
Chromosome Location
Chr.2: 93833467 - 93849995 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290782.1 NP_001277711.1 10 - 11 1109 76
NM_183220.3 NP_899043.1 10 - 11 1164 76
GenBank mRNA AK040509.1 - 7 - 8 2909 76
BC039569.1 - 10 - 11 1141 76

View Full Product Details