Assay Design



Gene Transcripts

Gene Symbol
Onecut2
Entrez Gene ID
225631
Gene Name
one cut domain, family member 2
Gene Aliases
C730009D12, OC-2, Oc2
Chromosome Location
Chr.18: 64340364 - 64398488 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194268.2 NP_919244.2 1 - 1 442 121
GenBank mRNA AY242995.1 - 1 - 1 442 121
BC103668.1 - 2 - 2 275 121

View Full Product Details