Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159904.1 | NP_001153376.1 | 2 - 3 | 336 | 145 |
NM_008527.2 | NP_032553.2 | 2 - 3 | 336 | 136 | |
GenBank mRNA | AF354263.1 | - | 2 - 3 | 197 | 145 |
AK079743.1 | - | 2 - 3 | 338 | 145 | |
BC061168.1 | - | 2 - 3 | 290 | 145 | |
M77678.1 | - | 2 - 3 | 275 | 136 |