Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010503.2 | NP_034633.2 | 1 - 1 | 65 | 158 |
GenBank mRNA | BC100688.1 | - | 1 - 1 | 158 | |
BC100689.1 | - | 1 - 1 | 158 | ||
BC100698.1 | - | 1 - 1 | 158 | ||
BC100699.1 | - | 1 - 1 | 158 | ||
BC114423.1 | - | 1 - 1 | 158 |