Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025329.3 | NP_079605.1 | 4 - 5 | 487 | 180 |
GenBank mRNA | AK002589.1 | - | 4 - 5 | 446 | 180 |
AK007178.1 | - | 4 - 5 | 475 | 180 | |
AK010027.1 | - | 4 - 5 | 445 | 180 | |
BC059716.1 | - | 4 - 5 | 488 | 180 |