Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025824.3 | NP_080100.1 | 4 - 5 | 427 | 150 |
GenBank mRNA | AK004784.2 | - | 4 - 5 | 427 | 150 |
AK013980.1 | - | 4 - 5 | 421 | 150 | |
AK029030.1 | - | 4 - 5 | 438 | 150 | |
AK076086.1 | - | 4 - 5 | 428 | 150 | |
AK076560.1 | - | 4 - 5 | 428 | 150 | |
AK087980.1 | - | 4 - 5 | 415 | 150 | |
AK088322.1 | - | 4 - 5 | 435 | 150 | |
AK133973.1 | - | 4 - 5 | 428 | 150 | |
AK151919.1 | - | 4 - 5 | 415 | 150 | |
AK152939.1 | - | 4 - 5 | 415 | 150 | |
AK153538.1 | - | 4 - 5 | 416 | 150 | |
AK161192.1 | - | 4 - 5 | 425 | 150 | |
AK166247.1 | - | 4 - 5 | 432 | 150 | |
AK167056.1 | - | 4 - 5 | 412 | 150 | |
AK167074.1 | - | 4 - 5 | 399 | 150 | |
AK167514.1 | - | 4 - 5 | 387 | 150 | |
AK167982.1 | - | 4 - 5 | 433 | 150 | |
AK168158.1 | - | 4 - 5 | 439 | 150 | |
BC005466.1 | - | 4 - 5 | 436 | 150 | |
BC028865.1 | - | 4 - 5 | 415 | 150 |