Assay Design



Gene Transcripts

Gene Symbol
Mif4gd
Entrez Gene ID
69674
Gene Name
MIF4G domain containing
Gene Aliases
1110014L05Rik, 2310075G12Rik
Chromosome Location
Chr.11: 115607918 - 115612973 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC026745.1 - 2 - 3 238 103
BC055812.1 - 1 - 2 604 103

View Full Product Details