Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029044.2 | NP_083320.1 | 3 - 4 | 348 | 169 |
GenBank mRNA | AK015430.1 | - | 3 - 4 | 350 | 169 |
AK047468.1 | - | 3 - 4 | 403 | 169 | |
AK049695.1 | - | 3 - 4 | 376 | 169 |