Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177460.4 | NP_803411.3 | 2 - 3 | 459 | 68 |
GenBank mRNA | AK033791.1 | - | - | 409 | 68 |
AK133475.1 | - | 2 - 3 | 460 | 68 | |
AK166002.1 | - | 1 - 2 | 223 | 68 | |
AK169866.1 | - | 2 - 3 | 374 | 68 | |
BC014731.1 | - | 1 - 2 | 489 | 68 | |
BC055447.1 | - | 2 - 3 | 428 | 68 |