Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009675.2 | NP_033805.1 | 3 - 4 | 2351 | 98 |
GenBank mRNA | AF054831.1 | - | 3 - 4 | 2366 | 98 |
AF115411.1 | - | 3 - 4 | 2182 | 98 | |
AK040729.1 | - | - | 1178 | 98 | |
BC080857.1 | - | 3 - 4 | 2197 | 98 |