Assay Design
Gene Transcripts
- Gene Symbol
- Gmcl1l
- Entrez Gene ID
-
71847
- Gene Name
- germ cell-less homolog 1 (Drosophila)-like
- Gene Aliases
-
1700017G21Rik, Gclh, Gmcl2
- Chromosome Location
-
Chr.X: 32560055 - 32562009 on Build GRCm38
- Gene Symbol
- Gm10921
- Entrez Gene ID
-
668963
- Gene Name
- predicted gene 10921
- Gene Aliases
-
ENSMUSG00000078215, OTTMUSG00000016420
- Chromosome Location
-
Chr.X: 4289286 - 4291245 on Build GRCm38
- Gene Symbol
- Gm10922
- Entrez Gene ID
-
668964
- Gene Name
- predicted gene 10922
- Gene Aliases
-
ENSMUSG00000078216, OTTMUSG00000016437
- Chromosome Location
-
Chr.X: 4370636 - 4372595 on Build GRCm38
- Gene Symbol
- Gm14345
- Entrez Gene ID
-
630022
- Gene Name
- predicted gene 14345
- Gene Aliases
-
OTTMUSG00000016405
- Chromosome Location
-
Chr.X: 3700233 - 3702192 on Build GRCm38
- Gene Symbol
- Gm14346
- Entrez Gene ID
-
668958
- Gene Name
- predicted gene 14346
- Gene Aliases
-
OTTMUSG00000016406
- Chromosome Location
-
Chr.X: 3441731 - 3443690 on Build GRCm38
- Gene Symbol
- Gm14351
- Entrez Gene ID
-
668960
- Gene Name
- predicted gene 14351
- Gene Aliases
-
OTTMUSG00000016411
- Chromosome Location
-
Chr.X: 3750931 - 3752885 on Build GRCm38
- Gene Symbol
- Gm21950
- Entrez Gene ID
-
100861966
- Gene Name
- predicted gene, 21950
- Gene Aliases
-
Mgclh
- Chromosome Location
-
Chr.X: 3076920 - 3078823 on Build GRCm38
- Gene Symbol
- Gm21951
- Entrez Gene ID
-
100862329
- Gene Name
- predicted gene, 21951
- Gene Aliases
-
Mgclh
- Chromosome Location
-
Chr.X: 32726137 - 32728057 on Build GRCm38
- Gene Symbol
- Gm2799
- Entrez Gene ID
-
100040482
- Gene Name
- predicted gene 2799
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 32278394 - 32280342 on Build GRCm38
- Gene Symbol
- Gm2825
- Entrez Gene ID
-
100040533
- Gene Name
- predicted gene 2825
- Gene Aliases
-
GMCL1P1, Gclh, Gmcl1l
- Chromosome Location
-
Chr.X: 32973897 - 32975846 on Build GRCm38
- Gene Symbol
- Gm2863
- Entrez Gene ID
-
100040606
- Gene Name
- predicted gene 2863
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 33313338 - 33315288 on Build GRCm38
- Gene Symbol
- Gm2913
- Entrez Gene ID
-
100040698
- Gene Name
- predicted gene 2913
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 33575387 - 33577346 on Build GRCm38
- Gene Symbol
- Gm2927
- Entrez Gene ID
-
100040722
- Gene Name
- predicted gene 2927
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 33657140 - 33659082 on Build GRCm38
- Gene Symbol
- Gm3701
- Entrez Gene ID
-
100042159
- Gene Name
- predicted gene 3701
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 3955043 - 3957002 on Build GRCm38
- Gene Symbol
- Gm3706
- Entrez Gene ID
-
100042168
- Gene Name
- predicted gene 3706
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 4037674 - 4039622 on Build GRCm38
- Gene Symbol
- Gm3750
- Entrez Gene ID
-
100042254
- Gene Name
- predicted gene 3750
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 4800411 - 4802359 on Build GRCm38
- Gene Symbol
- Gm3763
- Entrez Gene ID
-
100042275
- Gene Name
- predicted gene 3763
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 4952135 - 4954077 on Build GRCm38
- Gene Symbol
- Gm5925
- Entrez Gene ID
-
546185
- Gene Name
- germ cell-less homolog 1 family pseudogene
- Gene Aliases
-
EG546185
- Chromosome Location
-
Chr.X: 4515589 - 4517543 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_040410.1 |
|
1 - 1 |
1876 |
93 |
View Full Product Details