Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001081656.2 | NP_001075125.1 | 5 - 5 | 6085 | 128 |
GenBank mRNA | AK030658.1 | - | 1 - 1 | 1384 | 128 |
AK053917.1 | - | 1 - 1 | 2716 | 128 | |
AK082655.1 | - | 1 - 1 | 2767 | 128 |