Assay Design
Gene Transcripts
- Gene Symbol
- Mafa
- Entrez Gene ID
-
378435
- Gene Name
- v-maf musculoaponeurotic fibrosarcoma oncogene family, protein A (avian)
- Gene Aliases
-
RIPE3b1
- Chromosome Location
-
Chr.15: 75746843 - 75747922 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_194350.1 |
NP_919331.1 |
1 - 1 |
217 |
156 |
View Full Product Details