Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025394.3 | NP_079670.1 | 3 - 3 | 330 | 71 |
GenBank mRNA | AK003864.1 | - | 3 - 3 | 293 | 71 |
AK008581.1 | - | 3 - 3 | 247 | 71 | |
BC055352.1 | - | 3 - 3 | 308 | 71 | |
BE647192.1 | - | 2 - 2 | 132 | 71 | |
BU555283.1 | - | 3 - 3 | 333 | 71 | |
BY773505.1 | - | 3 - 3 | 323 | 71 |