Assay Design



Gene Transcripts

Gene Symbol
Olfr43
Entrez Gene ID
258706
Gene Name
olfactory receptor 43
Gene Aliases
IA7, MOR125-1, MOR125-5_p, Olfr403
Chromosome Location
Chr.11: 74206198 - 74207289 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146711.2 NP_666922.2 1 - 1 440 104
GenBank mRNA BC125464.1 - 1 - 1 440 104
BC125468.1 - 1 - 1 440 104

View Full Product Details