Assay Design
Gene Transcripts
- Gene Symbol
- Snrpd1
- Entrez Gene ID
-
20641
- Gene Name
- small nuclear ribonucleoprotein D1
- Gene Aliases
-
AA407109, AL023031, SMD1
- Chromosome Location
-
Chr.18: 10617796 - 10628230 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_009226.4 |
NP_033252.1 |
4 - 4 |
769 |
90 |
GenBank mRNA |
AK007674.1 |
- |
4 - 4 |
770 |
90 |
View Full Product Details