Assay Design



Gene Transcripts

Gene Symbol
Elovl6
Entrez Gene ID
170439
Gene Name
ELOVL family member 6, elongation of long chain fatty acids (yeast)
Gene Aliases
C77826, FAE, LCE
Chromosome Location
Chr.3: 129532386 - 129638495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130450.2 NP_569717.1 5 - 5 5768 149
GenBank mRNA AB072039.1 - 6 - 6 5675 149
AY053453.1 - 5 - 5 5732 149

View Full Product Details