Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019550.2 | NP_062423.1 | 13 - 14 | 1646 | 71 |
GenBank mRNA | AF095718.2 | - | 13 - 14 | 1516 | 71 |
AK135738.1 | - | 13 - 14 | 1649 | 71 | |
AK137348.1 | - | 13 - 14 | 1525 | 71 |