Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177323.3 | NP_796297.2 | 10 - 11 | 1561 | 122 |
GenBank mRNA | AK028707.1 | - | 9 - 10 | 1418 | 122 |
AK036805.1 | - | 10 - 11 | 1561 | 122 | |
AK133789.1 | - | 9 - 10 | 1661 | 122 | |
AK135334.1 | - | 10 - 11 | 1582 | 122 | |
AK144144.1 | - | 10 - 11 | 1584 | 122 | |
BC065059.1 | - | 10 - 11 | 1561 | 122 | |
BC145267.1 | - | 9 - 10 | 1377 | 122 | |
BC150745.1 | - | 9 - 10 | 1377 | 122 |