Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177751.2 | NP_808419.1 | 1 - 2 | 337 | 99 |
GenBank mRNA | AK122399.1 | - | 1 - 2 | 408 | 99 |
AK141544.1 | - | 1 - 2 | 408 | 99 | |
BC043093.1 | - | 1 - 2 | 337 | 99 | |
BC060716.1 | - | 1 - 2 | 337 | 99 |