Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016764.4 | NP_058044.1 | 5 - 6 | 831 | 96 |
GenBank mRNA | AK005031.1 | - | 5 - 6 | 810 | 96 |
AK015920.1 | - | 5 - 6 | 807 | 96 | |
AK146402.1 | - | 5 - 6 | 825 | 96 | |
AK152255.1 | - | 5 - 6 | 813 | 96 | |
BC003349.1 | - | 5 - 6 | 818 | 96 | |
BC019578.1 | - | 5 - 6 | 846 | 96 | |
U96746.1 | - | 5 - 6 | 829 | 96 |