Assay Design



Gene Transcripts

Gene Symbol
Phtf2
Entrez Gene ID
68770
Gene Name
putative homeodomain transcription factor 2
Gene Aliases
1110054G21Rik, 9530062N20Rik, AI447096, C77923
Chromosome Location
Chr.5: 20758664 - 20882124 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172992.3 NP_766580.3 17 - 18 2428 83
GenBank mRNA AK037021.1 - 17 - 18 2315 83
AK162226.1 - 6 - 7 873 83
BC052841.1 - 17 - 18 2341 83

View Full Product Details