Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021882.4 | NP_068682.2 | 10 - 11 | 1769 | 64 |
GenBank mRNA | AF119092.1 | - | 5 - 6 | 646 | 64 |
AK012808.1 | - | 10 - 11 | 1770 | 64 | |
AK131956.1 | - | 10 - 11 | 1771 | 64 | |
AK186888.1 | - | 2 - 3 | 290 | 64 | |
AK191933.1 | - | 2 - 3 | 290 | 64 | |
BC075667.1 | - | 10 - 11 | 1754 | 64 | |
BC082555.1 | - | 10 - 11 | 1755 | 64 |