Assay Design



Gene Transcripts

Gene Symbol
Agap3
Entrez Gene ID
213990
Gene Name
ArfGAP with GTPase domain, ankyrin repeat and PH domain 3
Gene Aliases
AGAP-3, AV028425, Centg3, Crag, Crag., MRIP-1, cnt-g3
Chromosome Location
Chr.5: 24452177 - 24502047 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256431.1 NP_001243360.1 7 - 8 995 84
NM_139153.2 NP_631892.2 7 - 8 995 84
GenBank mRNA AB078345.1 - 8 - 9 887 84
AF459091.1 - 7 - 8 995 84
AK079516.1 - 3 - 4 334 84

View Full Product Details