Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 3 - 4 | 505 | 153 |
GenBank mRNA | AB006036.1 | - | 3 - 4 | 298 | 153 |
AK014004.1 | - | 4 - 5 | 424 | 153 | |
AK136861.1 | - | 3 - 4 | 470 | 153 | |
AK161384.1 | - | 4 - 5 | 564 | 153 | |
AK169580.1 | - | 3 - 4 | 478 | 153 | |
BC020178.1 | - | 3 - 4 | 402 | 153 | |
BC062941.1 | - | 4 - 5 | 450 | 153 | |
U92456.1 | - | 4 - 5 | 427 | 153 |