Assay Design



Gene Transcripts

Gene Symbol
Dync1i1
Entrez Gene ID
13426
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DH IC-1, DHIC-1, DIC, Dncic1, IC74
Chromosome Location
Chr.6: 5725633 - 6028039 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191023.1 NP_001177952.1 9 - 10 1268 88
NM_001191025.1 NP_001177954.1 9 - 10 1217 88
NM_001191026.1 NP_001177955.1 10 - 11 1244 88
NM_001191027.1 NP_001177956.1 9 - 10 1184 88
NM_010063.4 NP_034193.2 10 - 11 1277 88
GenBank mRNA AF063229.1 - 10 - 11 1111 88
AF063230.1 - 9 - 10 1051 88
AK158589.1 - 10 - 11 1245 88
BC051992.1 - 10 - 11 1005 88
GU992206.1 - 9 - 10 1009 88
GU992207.1 - 9 - 10 925 88

View Full Product Details