Assay Design



Gene Transcripts

Gene Symbol
Dync1i1
Entrez Gene ID
13426
Gene Name
dynein cytoplasmic 1 intermediate chain 1
Gene Aliases
DH IC-1, DHIC-1, DIC, Dncic1, IC74
Chromosome Location
Chr.6: 5725633 - 6028039 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001191023.1 NP_001177952.1 15 - 16 2073 73
NM_001191025.1 NP_001177954.1 15 - 16 2022 73
NM_001191026.1 NP_001177955.1 16 - 17 2049 73
NM_001191027.1 NP_001177956.1 15 - 16 1989 73
NM_010063.4 NP_034193.2 16 - 17 2082 73
GenBank mRNA AF063229.1 - 16 - 17 73
AF063230.1 - 15 - 16 73
AK158589.1 - 16 - 17 2050 73
BC051992.1 - 16 - 17 1810 73
GU992206.1 - 15 - 16 1814 73
GU992207.1 - 15 - 16 1730 73

View Full Product Details